Order Entry
Puerto Rico
ContactUsLinkComponent
Human Bone Marrow Stromal Cell Antigen 1 ELISA Kit
Human Bone Marrow Stromal Cell Antigen 1 ELISA Kit
Catalog # 76740-976
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human Bone Marrow Stromal Cell Antigen 1 ELISA Kit
Catalog # 76740-976
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A77765-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Bone Marrow Stromal Cell Antigen 1
  • Description:
    Human Bone Marrow Stromal Cell antigen 1 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Human Bone Marrow Stromal Cell Antigen 1 ELISA Kit exhibits high specificity and excellent specificity for the detection of human Bone Marrow Stromal Cell Antigen 1. No significant cross-reactivity or interference between Bone Marrow Stromal Cell Antigen 1 and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.094 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76740-976

Specifications

About this item

Human Bone Marrow Stromal Cell Antigen 1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Bone Marrow Stromal Cell Antigen 1 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 0.156 to 10 ng/ml
  • Sensitivity: 0.094 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human Bone Marrow Stromal Cell Antigen 1 ELISA Kit (A77765) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Bone Marrow Stromal Cell Antigen 1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Bone Marrow Stromal Cell Antigen 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Bone Marrow Stromal Cell Antigen 1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Bone Marrow Stromal Cell Antigen 1 Antibody, which binds the captured Bone Marrow Stromal Cell Antigen 1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Bone Marrow Stromal Cell Antigen 1 captured in each well. The concentration of Bone Marrow Stromal Cell Antigen 1 can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.