Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human PIP5K2 gamma ELISA Kit
Catalog # 76740-876
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human PIP5K2 gamma ELISA Kit
Catalog # 76740-876
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A74942-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    PIP5K2 gamma
  • Description:
    Human PIP5K2 gamma ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Human PIP5K2 gamma ELISA kit exhibits high specificity and excellent specificity for the detection of human PIP5K2 gamma. No significant cross-reactivity or interference between PIP5K2 gamma and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    78.125 - 5000 pg/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    46.875 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76740-876

Specifications

About this item

Human PIP5K2 gamma ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PIP5K2 gamma in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 78.125 - 5000 pg/ml
  • Sensitivity: 46.875 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Human PIP5K2 gamma ELISA kit (A74942) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human PIP5K2 gamma in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PIP5K2 gamma has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PIP5K2 gamma present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PIP5K2 gamma Antibody, which binds the captured PIP5K2 gamma present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PIP5K2 gamma captured in each well. The concentration of PIP5K2 gamma can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.