Order Entry
Puerto Rico
ContactUsLinkComponent
Canine Adenosine A1 Receptor ELISA Kit
Canine Adenosine A1 Receptor ELISA Kit
Catalog # 76740-726
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Canine Adenosine A1 Receptor ELISA Kit
Catalog # 76740-726
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A74169-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Dog
  • Target protein:
    Adenosine A1 receptor
  • Description:
    Canine adenosine A1 receptor ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Canine Adenosine A1 receptor ELISA Kit exhibits high specificity and excellent specificity for the detection of canine Adenosine A1 receptor. No significant cross-reactivity or interference between Adenosine A1 receptor and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.094 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76740-726

Specifications

About this item

Canine Adenosine A1 receptor ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of canine Adenosine A1 receptor in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 0.156 to 10 ng/ml
  • Sensitivity: 0.094 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Canine Adenosine A1 Receptor ELISA Kit (A74169) employs the sandwich enzyme immunoassay technique for the quantitative measurement of canine Adenosine A1 Receptor in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Adenosine A1 Receptor has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Adenosine A1 Receptor present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Adenosine A1 Receptor Antibody, which binds the captured Adenosine A1 Receptor present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Adenosine A1 Receptor captured in each well. The concentration of Adenosine A1 Receptor can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.