Order Entry
Puerto Rico
ContactUsLinkComponent
Human DEFB119 ELISA Kit
Human DEFB119 ELISA Kit
Catalog # 76740-296
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human DEFB119 ELISA Kit
Catalog # 76740-296
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A74759-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    DEFB119
  • Description:
    Human DEFB119 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Human DEFB119 ELISA kit exhibits high specificity and excellent specificity for the detection of human DEFB119. No significant cross-reactivity or interference between DEFB119 and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.313 - 20 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    50 μl
  • Sensitivity:
    0.188 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76740-296

Specifications

About this item

Human DEFB119 ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of human DEFB119 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 0.313 - 20 ng/ml
  • Sensitivity: 0.188 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Human DEFB119 ELISA kit (A74759) employs the competitive enzyme immunoassay technique for the quantitative measurement of human DEFB119 in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with DEFB119 antigen. During the incubation, DEFB119 present in the samples or standards competes with the fixed amount of immobilized DEFB119 for binding sites on the Biotinylated Anti-DEFB119 Antibody. The more DEFB119 present in a sample or standard, the less Biotinylated Anti-DEFB119 Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-DEFB119 Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of DEFB119 present in each sample or standard. The concentration of DEFB119 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.