- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Procollagen Type I N-Terminal Propeptide/PINP
- Description:Rat procollagen type I N-terminal propeptide/PINP ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Rat Procollagen Type I N-Terminal Propeptide/PINP ELISA kit exhibits high specificity and excellent specificity for the detection of rat Procollagen Type I N-Terminal Propeptide/PINP. No significant cross-reactivity or interference between Procollagen Type I N-Terminal Propeptide/PINP and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:62.5 - 4000 pg/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:37.5 pg/ml
- Regulatory Status:RUO
- Cat. No.:76739-826
Rat Procollagen Type I N-Terminal Propeptide/PINP ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Procollagen Type I N-Terminal Propeptide/PINP in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 62.5 - 4000 pg/ml
- Sensitivity: 37.5 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Rat Procollagen Type I N-Terminal Propeptide/PINP ELISA kit (A79634) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Procollagen Type I N-Terminal Propeptide/PINP in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Procollagen Type I N-Terminal Propeptide/PINP has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Procollagen Type I N-Terminal Propeptide/PINP present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Procollagen Type I N-Terminal Propeptide/PINP Antibody, which binds the captured Procollagen Type I N-Terminal Propeptide/PINP present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Procollagen Type I N-Terminal Propeptide/PINP captured in each well. The concentration of Procollagen Type I N-Terminal Propeptide/PINP can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.