- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Urotensin II
- Description:Human Urotensin II ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Human Urotensin II ELISA Kit exhibits high specificity and excellent specificity for the detection of human Urotensin II. No significant cross-reactivity or interference between Urotensin II and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:15.625 - 1000 pg/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:9.375 pg/ml
- Regulatory Status:RUO
- Cat. No.:76739-546
Human Urotensin II ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Urotensin II in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 15.625 to 1000 pg/ml
- Sensitivity: 9.375 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Human Urotensin II ELISA Kit (A78953) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Urotensin II in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Urotensin II has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Urotensin II present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Urotensin II Antibody, which binds the captured Urotensin II present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Urotensin II captured in each well. The concentration of Urotensin II can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.