- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rabbit
- Target protein:N-Terminal Propeptide of Collagen alpha-1(I) Chain/PINP
- Description:Rabbit N-terminal propeptide of collagen alpha-1(I) chain/PINP ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross Reactivity:Rabbit N-Terminal Propeptide of Collagen alpha-1(I) Chain/PINP ELISA Kit exhibits high specificity and excellent specificity for the detection of rabbit N-Terminal Propeptide of Collagen alpha-1(I) Chain/PINP. No significant cross-reactivity or interference between N-Terminal Propeptide of Collagen alpha-1(I) Chain/PINP and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.156 - 10 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.094 ng/ml
- Regulatory Status:RUO
- Cat. No.:76738-906
Rabbit N-Terminal Propeptide of Collagen alpha-1(I) Chain/PINP ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rabbit N-Terminal Propeptide of Collagen alpha-1(I) Chain/PINP in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Rabbit N-Terminal Propeptide of Collagen alpha-1(I) Chain/PINP ELISA kit (A303849) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rabbit N-Terminal Propeptide of Collagen alpha-1(I) Chain/PINP in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for N-Terminal Propeptide of Collagen alpha-1(I) Chain/PINP has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the N-Terminal Propeptide of Collagen alpha-1(I) Chain/PINP present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-N-Terminal Propeptide of Collagen alpha-1(I) Chain/PINP Antibody, which binds the captured N-Terminal Propeptide of Collagen alpha-1(I) Chain/PINP present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of N-Terminal Propeptide of Collagen alpha-1(I) Chain/PINP captured in each well. The concentration of N-Terminal Propeptide of Collagen alpha-1(I) Chain/PINP can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.