Specifications
- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Calcium Channel L Type DHPR alpha 2 Subunit/CACNA2D1
- Description:Human Calcium channel L type DHPR alpha 2 subunit/CACNA2D1 ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross Reactivity:Human Calcium Channel L Type DHPR alpha 2 Subunit / CACNA2D1 ELISA Kit exhibits high specificity and excellent specificity for the detection of human Calcium Channel L Type DHPR alpha 2 Subunit / CACNA2D1. No significant cross-reactivity or interference between Calcium Channel L Type DHPR alpha 2 Subunit / CACNA2D1 and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:31.25 - 2000 pg/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:18.75 pg/ml
- Regulatory Status:RUO
- Cat. No.:76737-596
Specifications
About this item
Human Calcium Channel L Type DHPR alpha 2 Subunit/CACNA2D1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Calcium Channel L Type DHPR alpha 2 Subunit/CACNA2D1 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human Calcium Channel L Type DHPR alpha 2 Subunit/CACNA2D1 ELISA kit (A302960) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Calcium Channel L Type DHPR alpha 2 Subunit/CACNA2D1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Calcium Channel L Type DHPR alpha 2 Subunit/CACNA2D1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Calcium Channel L Type DHPR alpha 2 Subunit/CACNA2D1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated anti-Calcium Channel L Type DHPR alpha 2 Subunit/CACNA2D1 antibody, which binds the captured Calcium Channel L Type DHPR alpha 2 Subunit/CACNA2D1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Calcium Channel L Type DHPR alpha 2 Subunit/CACNA2D1 captured in each well. The concentration of Calcium Channel L Type DHPR alpha 2 Subunit/CACNA2D1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.