Order Entry
Puerto Rico
ContactUsLinkComponent
Human C4 Binding Protein/C4BPB ELISA Kit
Human C4 Binding Protein/C4BPB ELISA Kit
Catalog # 76735-326
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human C4 Binding Protein/C4BPB ELISA Kit
Catalog # 76735-326
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A76224-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    C4 Binding Protein/C4BPB
  • Description:
    Human C4 binding protein/C4BPB ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Human C4 Binding Protein/C4BPB ELISA Kit exhibits high specificity and excellent specificity for the detection of human C4 Binding Protein/C4BPB. No significant cross-reactivity or interference between C4 Binding Protein/C4BPB and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.469 - 30 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.281 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76735-326

Specifications

About this item

Human C4 Binding Protein/C4BPB ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human C4 Binding Protein/C4BPB in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 0.469 to 30 ng/ml
  • Sensitivity: 0.281 ng/ml
  • Sample type: Serum. plasma. tissue homogenates. and other biological fluids
  • Assay precision: Intra assay: CV <8%. Inter assay: CV <10%

Human C4 Binding Protein / C4BPB ELISA Kit (A76224) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human C4 Binding Protein / C4BPB in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for C4 Binding Protein / C4BPB has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the C4 Binding Protein / C4BPB present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-C4 Binding Protein / C4BPB Antibody, which binds the captured C4 Binding Protein / C4BPB present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of C4 Binding Protein / C4BPB captured in each well. The concentration of C4 Binding Protein / C4BPB can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.