- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Guinea pig
- Target protein:Adiponectin
- Description:Guinea pig adiponectin ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross Reactivity:Guinea Pig Adiponectin ELISA Kit exhibits high specificity and excellent specificity for the detection of guinea pig Adiponectin. No significant cross-reactivity or interference between Adiponectin and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:15.625 - 1000 pg/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:9.375 pg/ml
- Regulatory Status:RUO
- Cat. No.:76734-846
Guinea Pig Adiponectin ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of guinea pig Adiponectin in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Guinea Pig Adiponectin ELISA kit (A302599) employs the sandwich enzyme immunoassay technique for the quantitative measurement of guinea pig Adiponectin in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Adiponectin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Adiponectin present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated anti-Adiponectin antibody, which binds the captured Adiponectin present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Adiponectin captured in each well. The concentration of Adiponectin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.