Order Entry
Puerto Rico
ContactUsLinkComponent
Human C-terminal Propeptide of Collagen alpha-1(I) Chain/PICP ELISA Kit
Human C-terminal Propeptide of Collagen alpha-1(I) Chain/PICP ELISA Kit
Catalog # 76729-866
Supplier:  Antibodies.com
CAS Number:  
undefined
Human C-terminal Propeptide of Collagen alpha-1(I) Chain/PICP ELISA Kit
Catalog # 76729-866
Supplier:  Antibodies.com
Supplier Number:  A78625-96
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    PICP
  • Description:
    Human PICP ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Human C-terminal Propeptide of Collagen alpha-1(I) Chain/PICP ELISA Kit exhibits high specificity and excellent specificity for the detection of human C-terminal Propeptide of Collagen alpha-1(I) Chain/PICP. No significant cross-reactivity or interference between C-terminal Propeptide of Collagen alpha-1(I) Chain/PICP and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    62.5 - 4000 pg/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    37.5 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76729-866

Specifications

About this item

Human C-terminal Propeptide of Collagen alpha-1(I) Chain/PICP ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human C-terminal Propeptide of Collagen alpha-1(I) Chain/PICP in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 62.5 to 4000 pg/ml
  • Sensitivity: 37.5 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human C-terminal Propeptide of Collagen alpha-1(I) Chain / PICP ELISA Kit (A78625) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human C-terminal Propeptide of Collagen alpha-1(I) Chain / PICP in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for C-terminal Propeptide of Collagen alpha-1(I) Chain / PICP has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the C-terminal Propeptide of Collagen alpha-1(I) Chain / PICP present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-C-terminal Propeptide of Collagen alpha-1(I) Chain / PICP Antibody, which binds the captured C-terminal Propeptide of Collagen alpha-1(I) Chain / PICP present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of C-terminal Propeptide of Collagen alpha-1(I) Chain / PICP captured in each well. The concentration of C-terminal Propeptide of Collagen alpha-1(I) Chain / PICP can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.