Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Rabbit Progesterone ELISA Kit

Rabbit Progesterone ELISA Kit

Rabbit progesterone ELISA kit, 96 tests
Catalog # 76729-846
Supplier:   Antibodies.com
Supplier #:  A303856-96
Rabbit Progesterone ELISA Kit
Catalog # 76729-846
Supplier:  Antibodies.com
CAS Number:  
Rabbit Progesterone ELISA Kit
Rabbit Progesterone ELISA Kit

Rabbit progesterone ELISA kit, 96 tests

CAS Number:  
Price
$1,049.86
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Rabbit Progesterone ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of rabbit Progesterone in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Rabbit Progesterone ELISA kit (A303856) employs the competitive enzyme immunoassay technique for the quantitative measurement of rabbit Progesterone in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Progesterone antigen. During the incubation, Progesterone present in the samples or standards competes with the fixed amount of immobilized Progesterone for binding sites on the Biotinylated Anti-Progesterone Antibody. The more Progesterone present in a sample or standard, the less Biotinylated Anti-Progesterone Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Progesterone Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Progesterone present in each sample or standard. The concentration of Progesterone can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Description:
    Rabbit progesterone ELISA kit
  • Size:
    96 tests
  • Primary antibody reactivity:
    Rabbit
  • Detection Range:
    0.313 - 20 ng/ml
  • Sample Volume:
    50 μl
  • Time to Results:
    4 h 30 min
  • Detection Method:
    Colorimetric
  • Cat. No.:
    76729-846
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    0.188 ng/ml
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Competitive
  • Target protein:
    Progesterone
  • Sample Type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Shelf Life:
    Store for 6 months at 4 °C
  • Cross Reactivity:
    Rabbit Progesterone ELISA Kit exhibits high specificity and excellent specificity for the detection of rabbit Progesterone. No significant cross-reactivity or interference between Progesterone and analogues was observed.
  • Storage Temperature:
    4 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.