- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Monkey
- Target protein:Complement C3b
- Description:Monkey complement C3b ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross Reactivity:Monkey Complement C3b ELISA Kit exhibits high specificity and excellent specificity for the detection of monkey Complement C3b. No significant cross-reactivity or interference between Complement C3b and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.313 - 20 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.188 ng/ml
- Regulatory Status:RUO
- Cat. No.:76747-382
Monkey Complement C3b ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of monkey Complement C3b in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Detection Range: 0.313 to 20 ng/ml
- Sensitivity: 0.188 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%
Monkey Complement C3b ELISA kit (A303631) employs the sandwich enzyme immunoassay technique for the quantitative measurement of monkey Complement C3b in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Complement C3b has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Complement C3b present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated anti-Complement C3b antibody, which binds the captured Complement C3b present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Complement C3b captured in each well. The concentration of Complement C3b can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.