Order Entry
Puerto Rico
ContactUsLinkComponent
Rat Prostaglandin E Receptor EP2/PTGER2 ELISA Kit
Rat Prostaglandin E Receptor EP2/PTGER2 ELISA Kit
Catalog # 76747-172
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Rat Prostaglandin E Receptor EP2/PTGER2 ELISA Kit
Catalog # 76747-172
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A79972-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Prostaglandin E Receptor EP2/PTGER2
  • Description:
    Rat prostaglandin E receptor EP2/PTGER2 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Rat Prostaglandin E Receptor EP2/PTGER2 ELISA kit exhibits high specificity and excellent specificity for the detection of rat Prostaglandin E Receptor EP2/PTGER2. No significant cross-reactivity or interference between Prostaglandin E Receptor EP2/PTGER2 and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    39.063 - 2500 pg/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    23.438 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76747-172

Specifications

About this item

Rat Prostaglandin E Receptor EP2/PTGER2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Prostaglandin E Receptor EP2/PTGER2 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 39.063 - 2500 pg/ml
  • Sensitivity: 23.438 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Rat Prostaglandin E Receptor EP2/PTGER2 ELISA kit (A79972) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Prostaglandin E Receptor EP2/PTGER2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Prostaglandin E Receptor EP2/PTGER2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Prostaglandin E Receptor EP2/PTGER2 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Prostaglandin E Receptor EP2/PTGER2 Antibody, which binds the captured Prostaglandin E Receptor EP2/PTGER2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Prostaglandin E Receptor EP2/PTGER2 captured in each well. The concentration of Prostaglandin E Receptor EP2/PTGER2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.