- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Complement Factor I
- Description:Human complement factor I ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Human Complement Factor I ELISA Kit exhibits high specificity and excellent specificity for the detection of human Complement Factor I. No significant cross-reactivity or interference between Complement Factor I and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:93.75 - 6000 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:56.25 ng/ml
- Regulatory Status:RUO
- Cat. No.:76747-112
Human Complement Factor I ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Complement Factor I in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA kit
- Detection Range: 93.75 to 6000 ng/ml
- Sensitivity: 56.25 ng/ml
- Sample Type: Serum, plasma, tissue homogenates and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Human Complement Factor I ELISA kit (A79200) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Complement Factor I in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Complement Factor I has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Complement Factor I present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Complement Factor I Antibody, which binds the captured Complement Factor I present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Complement Factor I captured in each well. The concentration of Complement Factor I can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.