- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Pulmonary Surfactant-Associated Protein B
- Description:Human Pulmonary Surfactant-Associated protein B ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Human Pulmonary Surfactant-Associated Protein B ELISA Kit exhibits high specificity and excellent specificity for the detection of human Pulmonary Surfactant-Associated Protein B. No significant cross-reactivity or interference between Pulmonary Surfactant-Associated Protein B and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.781 - 50 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.469 ng/ml
- Regulatory Status:RUO
- Cat. No.:76747-032
Human Pulmonary Surfactant-Associated Protein B ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Pulmonary Surfactant-Associated Protein B in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 0.781 to 50 ng/ml
- Sensitivity: 0.469 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Human Pulmonary Surfactant-Associated Protein B ELISA Kit (A77363) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Pulmonary Surfactant-Associated Protein B in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Pulmonary Surfactant-Associated Protein B has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Pulmonary Surfactant-Associated Protein B present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Pulmonary Surfactant-Associated Protein B Antibody, which binds the captured Pulmonary Surfactant-Associated Protein B present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Pulmonary Surfactant-Associated Protein B captured in each well. The concentration of Pulmonary Surfactant-Associated Protein B can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.