Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Cortisol Binding Globulin ELISA Kit
Mouse Cortisol Binding Globulin ELISA Kit
Catalog # 76744-332
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse Cortisol Binding Globulin ELISA Kit
Catalog # 76744-332
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A74539-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Cortisol Binding Globulin
  • Description:
    Mouse cortisol binding globulin ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Mouse Cortisol Binding Globulin ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Cortisol Binding Globulin. No significant cross-reactivity or interference between Cortisol Binding Globulin and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    15.625 - 1000 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    9.375 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76744-332

Specifications

About this item

Mouse Cortisol Binding Globulin ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Cortisol Binding Globulin in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 15.625 to 1000 ng/ml
  • Sensitivity: 9.375 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Mouse Cortisol Binding Globulin ELISA Kit (A74539) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Cortisol Binding Globulin in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Cortisol Binding Globulin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Cortisol Binding Globulin present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Cortisol Binding Globulin Antibody, which binds the captured Cortisol Binding Globulin present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Cortisol Binding Globulin captured in each well. The concentration of Cortisol Binding Globulin can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.