- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Myelin Oligodendrocyte Glycoprotein
- Description:Human myelin oligodendrocyte glycoprotein ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Human Myelin Oligodendrocyte Glycoprotein ELISA Kit exhibits high specificity and excellent specificity for the detection of human Myelin Oligodendrocyte Glycoprotein. No significant cross-reactivity or interference between Myelin Oligodendrocyte Glycoprotein and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.156 - 10 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.094 ng/ml
- Regulatory Status:RUO
- Cat. No.:76743-532
Human Myelin Oligodendrocyte Glycoprotein ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Myelin Oligodendrocyte Glycoprotein in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA kit
- Detection Range: 0.156 to 10 ng/ml
- Sensitivity: 0.094 ng/ml
- Sample Type: Serum, plasma, tissue homogenates and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Human Myelin Oligodendrocyte Glycoprotein ELISA kit (A79540) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Myelin Oligodendrocyte Glycoprotein in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Myelin Oligodendrocyte Glycoprotein has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Myelin Oligodendrocyte Glycoprotein present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Myelin Oligodendrocyte Glycoprotein Antibody, which binds the captured Myelin Oligodendrocyte Glycoprotein present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Myelin Oligodendrocyte Glycoprotein captured in each well. The concentration of Myelin Oligodendrocyte Glycoprotein can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.