Order Entry
Puerto Rico
ContactUsLinkComponent
Rat Pituitary Adenylate Cyclase-Activating Polypeptide ELISA Kit
Rat Pituitary Adenylate Cyclase-Activating Polypeptide ELISA Kit
Catalog # 76743-432
Supplier:  Antibodies.com
CAS Number:  
undefined
Rat Pituitary Adenylate Cyclase-Activating Polypeptide ELISA Kit
Catalog # 76743-432
Supplier:  Antibodies.com
Supplier Number:  A76106-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Pituitary Adenylate Cyclase-Activating Polypeptide
  • Description:
    Rat pituitary adenylate cyclase-activating polypeptide ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Rat Pituitary Adenylate Cyclase-Activating Polypeptide ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Pituitary Adenylate Cyclase-Activating Polypeptide. No significant cross-reactivity or interference between Pituitary Adenylate Cyclase-Activating Polypeptide and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    6.25 - 400 pg/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    3.75 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76743-432

Specifications

About this item

Rat Pituitary Adenylate Cyclase-Activating Polypeptide ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Pituitary Adenylate Cyclase-Activating Polypeptide in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 6.25 to 400 pg/ml
  • Sensitivity: 3.75 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat Pituitary Adenylate Cyclase-Activating Polypeptide ELISA Kit (A76106) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Pituitary Adenylate Cyclase-Activating Polypeptide in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Pituitary Adenylate Cyclase-Activating Polypeptide has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Pituitary Adenylate Cyclase-Activating Polypeptide present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Pituitary Adenylate Cyclase-Activating Polypeptide Antibody, which binds the captured Pituitary Adenylate Cyclase-Activating Polypeptide present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Pituitary Adenylate Cyclase-Activating Polypeptide captured in each well. The concentration of Pituitary Adenylate Cyclase-Activating Polypeptide can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.