- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Adenosine A1 Receptor
- Description:Mouse adenosine A1 receptor ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross Reactivity:Mouse Adenosine A1 Receptor ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Adenosine A1 Receptor. No significant cross-reactivity or interference between Adenosine A1 Receptor and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:78.125 - 5000 pg/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:46.875 pg/ml
- Regulatory Status:RUO
- Cat. No.:76743-252
Mouse Adenosine A1 receptor ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Adenosine A1 receptor in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Mouse Adenosine A1 receptor ELISA kit (A303563) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Adenosine A1 receptor in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Adenosine A1 receptor has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Adenosine A1 receptor present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated anti-Adenosine A1 receptor antibody, which binds the captured Adenosine A1 receptor present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Adenosine A1 receptor captured in each well. The concentration of Adenosine A1 receptor can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.