- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:PARM-1
- Description:Mouse PARM-1 ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Mouse PARM-1 ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse PARM-1. No significant cross-reactivity or interference between PARM-1 and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.781 - 50 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.469 ng/ml
- Regulatory Status:RUO
- Cat. No.:76742-472
Mouse PARM-1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse PARM-1 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 0.781 to 50 ng/ml
- Sensitivity: 0.469 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Mouse PARM-1 ELISA Kit (A77092) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse PARM-1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PARM-1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PARM-1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PARM-1 Antibody, which binds the captured PARM-1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PARM-1 captured in each well. The concentration of PARM-1 can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.