Order Entry
Puerto Rico
ContactUsLinkComponent
Human FSTL1/FRP ELISA Kit
Human FSTL1/FRP ELISA Kit
Catalog # 76741-702
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human FSTL1/FRP ELISA Kit
Catalog # 76741-702
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A76574-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    FSTL1/FRP
  • Description:
    Human FSTL1/FRP ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Human FSTL1/FRP ELISA kit exhibits high specificity and excellent specificity for the detection of human FSTL1/FRP. No significant cross-reactivity or interference between FSTL1/FRP and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.313 - 20 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.188 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76741-702

Specifications

About this item

Human FSTL1/FRP ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human FSTL1/FRP in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 0.313 to 20 ng/ml
  • Sensitivity: 0.188 ng/ml
  • Sample type: Serum. plasma. tissue homogenates. and other biological fluids
  • Assay precision: Intra assay: CV <8%. Inter assay: CV <10%

Human FSTL1/FRP ELISA kit (A76574) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human FSTL1/FRP in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for FSTL1/FRP has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the FSTL1/FRP present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-FSTL1/FRP Antibody, which binds the captured FSTL1/FRP present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of FSTL1/FRP captured in each well. The concentration of FSTL1/FRP can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.