- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:GOT1L1
- Description:Human GOT1L1 ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Human GOT1L1 ELISA kit exhibits high specificity and excellent specificity for the detection of human GOT1L1. No significant cross-reactivity or interference between GOT1L1 and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:3.906 - 250 pg/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:2.344 pg/ml
- Regulatory Status:RUO
- Cat. No.:76741-572
Human GOT1L1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human GOT1L1 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 3.906 - 250 pg/ml
- Sensitivity: 2.344 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Human GOT1L1 ELISA kit (A75445) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human GOT1L1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for GOT1L1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the GOT1L1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-GOT1L1 Antibody, which binds the captured GOT1L1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of GOT1L1 captured in each well. The concentration of GOT1L1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.