Order Entry
Puerto Rico
ContactUsLinkComponent
Human Glutathione S Transferase alpha 1 ELISA Kit
Human Glutathione S Transferase alpha 1 ELISA Kit
Catalog # 76740-562
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human Glutathione S Transferase alpha 1 ELISA Kit
Catalog # 76740-562
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A75455-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Glutathione S Transferase alpha 1
  • Description:
    Human glutathione S transferase alpha 1 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Human Glutathione S Transferase alpha 1 ELISA kit exhibits high specificity and excellent specificity for the detection of human Glutathione S Transferase alpha 1. No significant cross-reactivity or interference between Glutathione S Transferase alpha 1 and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.625 - 40 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.375 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76740-562

Specifications

About this item

Human Glutathione S Transferase alpha 1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Glutathione S Transferase alpha 1 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 0.625 - 40 ng/ml
  • Sensitivity: 0.375 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Human Glutathione S Transferase alpha 1 ELISA kit (A75455) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Glutathione S Transferase alpha 1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Glutathione S Transferase alpha 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Glutathione S Transferase alpha 1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Glutathione S Transferase alpha 1 Antibody, which binds the captured Glutathione S Transferase alpha 1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Glutathione S Transferase alpha 1 captured in each well. The concentration of Glutathione S Transferase alpha 1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.