Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse C-Type Natriuretic Peptide ELISA Kit
Mouse C-Type Natriuretic Peptide ELISA Kit
Catalog # 76740-502
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse C-Type Natriuretic Peptide ELISA Kit
Catalog # 76740-502
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A78536-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    C-Type Natriuretic Peptide
  • Description:
    Mouse C-type natriuretic peptide ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Mouse C-Type Natriuretic Peptide ELISA kit exhibits high specificity and excellent specificity for the detection of mouse C-Type Natriuretic Peptide. No significant cross-reactivity or interference between C-Type Natriuretic Peptide and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    15.625 - 1000 pg/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    50 μl
  • Sensitivity:
    9.375 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76740-502

Specifications

About this item

Mouse C-Type Natriuretic Peptide ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of mouse C-Type Natriuretic Peptide in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 15.625 to 1000 pg/ml
  • Sensitivity: 9.375 pg/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse C-Type Natriuretic Peptide ELISA kit (A78536) employs the competitive enzyme immunoassay technique for the quantitative measurement of mouse C-Type Natriuretic Peptide in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with C-Type Natriuretic Peptide antigen. During the incubation, C-Type Natriuretic Peptide present in the samples or standards competes with the fixed amount of immobilized C-Type Natriuretic Peptide for binding sites on the Biotinylated Anti-C-Type Natriuretic Peptide Antibody. The more C-Type Natriuretic Peptide present in a sample or standard, the less Biotinylated Anti-C-Type Natriuretic Peptide Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-C-Type Natriuretic Peptide Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of C-Type Natriuretic Peptide present in each sample or standard. The concentration of C-Type Natriuretic Peptide can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.