- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Guinea pig
- Target protein:beta Nerve Growth Factor
- Description:Guinea pig beta nerve growth factor ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Guinea pig beta Nerve Growth Factor ELISA Kit exhibits high specificity and excellent specificity for the detection of guinea pig beta Nerve Growth Factor. No significant cross-reactivity or interference between beta Nerve Growth Factor and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:15.625 - 1000 pg/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:9.375 pg/ml
- Regulatory Status:RUO
- Cat. No.:76740-092
Guinea pig beta Nerve Growth Factor ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of guinea pig beta Nerve Growth Factor in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 15.625 to 1000 pg/ml
- Sensitivity: 9.375 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Guinea Pig beta Nerve Growth Factor ELISA Kit (A74100) employs the sandwich enzyme immunoassay technique for the quantitative measurement of guinea pig beta Nerve Growth Factor in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for beta Nerve Growth Factor has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the beta Nerve Growth Factor present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-beta Nerve Growth Factor Antibody, which binds the captured beta Nerve Growth Factor present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of beta Nerve Growth Factor captured in each well. The concentration of beta Nerve Growth Factor can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.