- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Estrogen Related Receptor gamma
- Description:Rat estrogen related receptor gamma ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Rat Estrogen Related Receptor gamma ELISA kit exhibits high specificity and excellent specificity for the detection of rat Estrogen Related Receptor gamma. No significant cross-reactivity or interference between Estrogen Related Receptor gamma and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.313 - 20 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.188 ng/ml
- Regulatory Status:RUO
- Cat. No.:76740-052
Rat Estrogen Related Receptor gamma ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Estrogen Related Receptor gamma in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 0.313 - 20 ng/ml
- Sensitivity: 0.188 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Rat Estrogen Related Receptor gamma ELISA kit (A79303) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Estrogen Related Receptor gamma in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Estrogen Related Receptor gamma has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Estrogen Related Receptor gamma present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Estrogen Related Receptor gamma Antibody, which binds the captured Estrogen Related Receptor gamma present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Estrogen Related Receptor gamma captured in each well. The concentration of Estrogen Related Receptor gamma can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.