Order Entry
Puerto Rico
ContactUsLinkComponent
Human Selenium Binding Protein 1/SBP ELISA Kit
Human Selenium Binding Protein 1/SBP ELISA Kit
Catalog # 76737-602
Supplier:  Antibodies.com
CAS Number:  
undefined
Human Selenium Binding Protein 1/SBP ELISA Kit
Catalog # 76737-602
Supplier:  Antibodies.com
Supplier Number:  A74535-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Selenium Binding Protein 1/SBP
  • Description:
    Human selenium binding protein 1/SBP ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Human Selenium Binding Protein 1/SBP ELISA kit exhibits high specificity and excellent specificity for the detection of human Selenium Binding Protein 1/SBP. No significant cross-reactivity or interference between Selenium Binding Protein 1/SBP and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    31.25 - 2000 pg/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    18.75 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76737-602

Specifications

About this item

Human Selenium Binding Protein 1/SBP ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Selenium Binding Protein 1/SBP in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 31.25 - 2000 pg/ml
  • Sensitivity: 18.75 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Human Selenium Binding Protein 1/SBP ELISA kit (A74535) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Selenium Binding Protein 1/SBP in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Selenium Binding Protein 1/SBP has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Selenium Binding Protein 1/SBP present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Selenium Binding Protein 1/SBP Antibody, which binds the captured Selenium Binding Protein 1/SBP present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Selenium Binding Protein 1/SBP captured in each well. The concentration of Selenium Binding Protein 1/SBP can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.