- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Guinea pig
- Target protein:Cross Linked C-telopeptide of type III Collagen/CTXIII
- Description:Guinea pig cross linked C-telopeptide of type III collagen/CTXIII ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Guinea pig Cross Linked C-telopeptide of type III Collagen/CTXIII ELISA Kit exhibits high specificity and excellent specificity for the detection of guinea pig Cross Linked C-telopeptide of type III Collagen/CTXIII. No significant cross-reactivity or interference between Cross Linked C-telopeptide of type III Collagen/CTXIII and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.156 - 10 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.094 ng/ml
- Regulatory Status:RUO
- Cat. No.:76737-322
Guinea pig Cross Linked C-telopeptide of type III Collagen/CTXIII ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of guinea pig Cross Linked C-telopeptide of type III Collagen/CTXIII in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 0.156 to 10 ng/ml
- Sensitivity: 0.094 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Guinea Pig Cross Linked C-telopeptide of Type III Collagen / CTXIII ELISA Kit (A73812) employs the sandwich enzyme immunoassay technique for the quantitative measurement of guinea pig Cross Linked C-telopeptide of Type III Collagen / CTXIII in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Cross Linked C-telopeptide of Type III Collagen / CTXIII has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Cross Linked C-telopeptide of Type III Collagen / CTXIII present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Cross Linked C-telopeptide of Type III Collagen / CTXIII Antibody, which binds the captured Cross Linked C-telopeptide of Type III Collagen / CTXIII present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Cross Linked C-telopeptide of Type III Collagen / CTXIII captured in each well. The concentration of Cross Linked C-telopeptide of Type III Collagen / CTXIII can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.