Order Entry
Puerto Rico
ContactUsLinkComponent
Human APOA4/Apo-AIV ELISA Kit
Human APOA4/Apo-AIV ELISA Kit
Catalog # 76735-522
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human APOA4/Apo-AIV ELISA Kit
Catalog # 76735-522
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A76154-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    APOA4/Apo-AIV
  • Description:
    Human APOA4/Apo-AIV ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Human APOA4/Apo-AIV ELISA Kit exhibits high specificity and excellent specificity for the detection of human APOA4/Apo-AIV. No significant cross-reactivity or interference between APOA4/Apo-AIV and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    12.5 - 800 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    7.5 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76735-522

Specifications

About this item

Human APOA4/Apo-AIV ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human APOA4/Apo-AIV in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 12.5 to 800 ng/ml
  • Sensitivity: 7.5 ng/ml
  • Sample type: Serum. plasma. tissue homogenates and other biological fluids
  • Assay precision: Intra assay: CV <8%. Inter assay: CV <10%

Human APOA4/Apo-AIV ELISA kit (A76154) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human APOA4/Apo-AIV in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for APOA4/Apo-AIV has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the APOA4/Apo-AIV present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-APOA4/Apo-AIV Antibody, which binds the captured APOA4/Apo-AIV present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of APOA4/Apo-AIV captured in each well. The concentration of APOA4/Apo-AIV can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.