Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Human SMC4 ELISA Kit

Human SMC4 ELISA kit, 96 tests
Catalog # 76735-012
Supplier:   Antibodies.com
 :  A78803-96
Human SMC4 ELISA Kit
Catalog # 76735-012
Supplier:  Antibodies.com
CAS Number:  
Human SMC4 ELISA Kit
Human SMC4 ELISA Kit

Human SMC4 ELISA kit, 96 tests

CAS Number:  
Price
$677.87
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Human SMC4 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human SMC4 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 78.125 to 5000 pg/ml
  • Sensitivity: 46.875 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human SMC4 ELISA Kit (A78803) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human SMC4 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for SMC4 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SMC4 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-SMC4 Antibody, which binds the captured SMC4 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of SMC4 captured in each well. The concentration of SMC4 can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

 : This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Description:
    Human SMC4 ELISA kit
  • Size:
    96 tests
  • Primary antibody reactivity:
    Human
  • Detection Range:
    78.125 - 5000 pg/ml
  • Sample Volume:
    100 μl
  • Time to Results:
    4 h 30 min
  • Detection Method:
    Colorimetric
  • Assay Principle:
    Quantitative
  • Cat. No.:
    76735-012
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    46.875 pg/ml
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Sandwich
  • Target protein:
    SMC4
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Shelf Life:
    Store for 6 months at 4 °C
  • Cross Reactivity:
    Human SMC4 ELISA Kit exhibits high specificity and excellent specificity for the detection of human SMC4. No significant cross-reactivity or interference between SMC4 and analogues was observed.
  • Storage Temperature:
    4 °C

Specifications