Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Monkey Soluble CD163 ELISA Kit

Monkey Soluble CD163 ELISA Kit

Monkey soluble CD163 ELISA kit, 96 tests
Catalog # 76734-942
Supplier:   Antibodies.com
Supplier #:  A74591-96
Monkey Soluble CD163 ELISA Kit
Catalog # 76734-942
Supplier:  Antibodies.com
CAS Number:  
Monkey Soluble CD163 ELISA Kit
Monkey Soluble CD163 ELISA Kit

Monkey soluble CD163 ELISA kit, 96 tests

CAS Number:  
Price
$789.97
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Monkey Soluble CD163 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of monkey Soluble CD163 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 0.781 to 50 ng/ml
  • Sensitivity: 0.469 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Monkey Soluble CD163 ELISA Kit (A74591) employs the sandwich enzyme immunoassay technique for the quantitative measurement of monkey Soluble CD163 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Soluble CD163 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Soluble CD163 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Soluble CD163 Antibody, which binds the captured Soluble CD163 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Soluble CD163 captured in each well. The concentration of Soluble CD163 can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

 : This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Description:
    Monkey soluble CD163 ELISA kit
  • Size:
    96 tests
  • Primary antibody reactivity:
    Monkey
  • Detection Range:
    0.781 - 50 ng/ml
  • Sample Volume:
    100 μl
  • Time to Results:
    4 h 30 min
  • Detection Method:
    Colorimetric
  • Assay Principle:
    Quantitative
  • Cat. No.:
    76734-942
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    0.469 ng/ml
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Sandwich
  • Target protein:
    Soluble CD163
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Shelf Life:
    Store for 6 months at 4 °C
  • Cross Reactivity:
    Monkey Soluble CD163 ELISA Kit exhibits high specificity and excellent specificity for the detection of monkey Soluble CD163. No significant cross-reactivity or interference between Soluble CD163 and analogues was observed.
  • Storage Temperature:
    4 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.