Order Entry
Puerto Rico
ContactUsLinkComponent
Canine CCL4/MIP-1 Beta ELISA Kit
Canine CCL4/MIP-1 Beta ELISA Kit
Catalog # 76734-102
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Canine CCL4/MIP-1 Beta ELISA Kit
Catalog # 76734-102
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A302573-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Dog
  • Target protein:
    CCL4/MIP-1 beta
  • Description:
    Canine CCL4/MIP-1 beta ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross Reactivity:
    Canine CCL4/MIP-1 beta ELISA Kit exhibits high specificity and excellent specificity for the detection of canine CCL4/MIP-1 beta. No significant cross-reactivity or interference between CCL4/MIP-1 beta and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    7.813 - 500 pg/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    4.688 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76734-102

Specifications

About this item

Canine CCL4/MIP-1 beta ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of canine CCL4/MIP-1 beta in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Detection Range: 7.813 - 500 pg/ml
  • Sensitivity: 4.688 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Canine CCL4/MIP-1 beta ELISA kit (A302573) employs the sandwich enzyme immunoassay technique for the quantitative measurement of canine CCL4/MIP-1 beta in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for CCL4/MIP-1 beta has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the CCL4/MIP-1 beta present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated anti-CCL4/MIP-1 beta antibody, which binds the captured CCL4/MIP-1 beta present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of CCL4/MIP-1 beta captured in each well. The concentration of CCL4/MIP-1 beta can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.