- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Vitamin D Binding Protein
- Description:Rat Vitamin D binding protein ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Rat Vitamin D Binding Protein ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Vitamin D Binding Protein. No significant cross-reactivity or interference between Vitamin D Binding Protein and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:15.625 - 1000 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:9.375 ng/ml
- Regulatory Status:RUO
- Cat. No.:76732-702
Rat Vitamin D Binding Protein ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Vitamin D Binding Protein in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 15.625 to 1000 ng/ml
- Sensitivity: 9.375 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Rat Vitamin D Binding Protein ELISA Kit (A79960) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Vitamin D Binding Protein in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Vitamin D Binding Protein has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Vitamin D Binding Protein present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Vitamin D Binding Protein Antibody, which binds the captured Vitamin D Binding Protein present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Vitamin D Binding Protein captured in each well. The concentration of Vitamin D Binding Protein can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.