- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Des-gamma
- Description:Rat Des-gamma carboxyprothrombin ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Rat Des-gamma Carboxyprothrombin ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Des-gamma Carboxyprothrombin. No significant cross-reactivity or interference between Des-gamma Carboxyprothrombin and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.156 - 10 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.094 ng/ml
- Regulatory Status:RUO
- Cat. No.:76732-182
Rat Des-gamma Carboxyprothrombin ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Des-gamma Carboxyprothrombin in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 0.156 to 10 ng/ml
- Sensitivity: 0.094 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Rat Des-gamma Carboxyprothrombin ELISA Kit (A80284) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Des-gamma Carboxyprothrombin in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Des-gamma Carboxyprothrombin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Des-gamma Carboxyprothrombin present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Des-gamma Carboxyprothrombin Antibody, which binds the captured Des-gamma Carboxyprothrombin present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Des-gamma Carboxyprothrombin captured in each well. The concentration of Des-gamma Carboxyprothrombin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.