- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Competitive
- Format:Pre-coated
- Primary antibody reactivity:Monkey
- Target protein:Luteinizing Hormone
- Description:Monkey luteinizing hormone ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross Reactivity:Monkey Luteinizing Hormone ELISA Kit exhibits high specificity and excellent specificity for the detection of monkey Luteinizing Hormone. No significant cross-reactivity or interference between Luteinizing Hormone and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.938 - 60 mIU/ml
- Storage Temperature:4 °C
- Sample Volume:50 μl
- Sensitivity:0.563 mIU/ml
- Regulatory Status:RUO
- Cat. No.:76744-444
Monkey Luteinizing Hormone ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of monkey Luteinizing Hormone in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Monkey Luteinizing Hormone ELISA kit (A303637) employs the competitive enzyme immunoassay technique for the quantitative measurement of monkey Luteinizing Hormone in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Luteinizing Hormone antigen. During the incubation, Luteinizing Hormone present in the samples or standards competes with the fixed amount of immobilized Luteinizing Hormone for binding sites on the Biotinylated Anti-Luteinizing Hormone Antibody. The more Luteinizing Hormone present in a sample or standard, the less Biotinylated Anti-Luteinizing Hormone Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Luteinizing Hormone Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Luteinizing Hormone present in each sample or standard. The concentration of Luteinizing Hormone can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.