Order Entry
Puerto Rico
ContactUsLinkComponent
Rat Telomerase Reverse Transcriptase ELISA Kit
Rat Telomerase Reverse Transcriptase ELISA Kit
Catalog # 76742-654
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Rat Telomerase Reverse Transcriptase ELISA Kit
Catalog # 76742-654
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A247110-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Telomerase Reverse Transcriptase
  • Description:
    Rat telomerase reverse transcriptase ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross Reactivity:
    Rat Telomerase Reverse Transcriptase ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Telomerase Reverse Transcriptase. No significant cross-reactivity or interference between Telomerase Reverse Transcriptase and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.094 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76742-654

Specifications

About this item

Rat Telomerase Reverse Transcriptase ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Telomerase Reverse Transcriptase in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Rat Telomerase Reverse Transcriptase ELISA kit (A247110) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Telomerase Reverse Transcriptase in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Telomerase Reverse Transcriptase has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Telomerase Reverse Transcriptase present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Telomerase Reverse Transcriptase Antibody, which binds the captured Telomerase Reverse Transcriptase present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Telomerase Reverse Transcriptase captured in each well. The concentration of Telomerase Reverse Transcriptase can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.