Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse alpha Hemoglobin Stabilizing Protein ELISA Kit
Mouse alpha Hemoglobin Stabilizing Protein ELISA Kit
Catalog # 76742-064
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse alpha Hemoglobin Stabilizing Protein ELISA Kit
Catalog # 76742-064
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A77656-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    alpha Hemoglobin Stabilizing Protein
  • Description:
    Mouse alpha hemoglobin stabilizing protein ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Mouse alpha Hemoglobin Stabilizing Protein ELISA kit exhibits high specificity and excellent specificity for the detection of mouse alpha Hemoglobin Stabilizing Protein. No significant cross-reactivity or interference between alpha Hemoglobin Stabilizing Protein and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.313 - 20 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.188 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76742-064

Specifications

About this item

Mouse alpha Hemoglobin Stabilizing Protein ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse alpha Hemoglobin Stabilizing Protein in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 0.313 - 20 ng/ml
  • Sensitivity: 0.188 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Mouse alpha Hemoglobin Stabilizing Protein ELISA kit (A77656) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse alpha Hemoglobin Stabilizing Protein in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for alpha Hemoglobin Stabilizing Protein has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the alpha Hemoglobin Stabilizing Protein present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-alpha Hemoglobin Stabilizing Protein Antibody, which binds the captured alpha Hemoglobin Stabilizing Protein present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of alpha Hemoglobin Stabilizing Protein captured in each well. The concentration of alpha Hemoglobin Stabilizing Protein can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.