Specifications
- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Alkaline phosphatase, Tissue Non-Specific
- Description:Rat alkaline phosphatase, tissue non-specific ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Rat Alkaline Phosphatase, Tissue Non-Specific ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Alkaline Phosphatase, Tissue Non-Specific. No significant cross-reactivity or interference between Alkaline Phosphatase, Tissue Non-Specific and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.156 - 10 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.094 ng/ml
- Regulatory Status:RUO
- Cat. No.:76740-714
Specifications
About this item
Rat Alkaline Phosphatase, Tissue Non-Specific ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Alkaline Phosphatase, Tissue Non-Specific in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA kit
- Detection Range: 0.156 to 10 ng/ml
- Sensitivity: 0.094 ng/ml
- Sample Type: Serum, plasma, tissue homogenates and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Rat Alkaline Phosphatase, Tissue Non-Specific ELISA kit (A79131) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Alkaline Phosphatase, Tissue Non-Specific in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Alkaline Phosphatase, Tissue Non-Specific has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Alkaline Phosphatase, Tissue Non-Specific present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Alkaline Phosphatase, Tissue Non-Specific Antibody, which binds the captured Alkaline Phosphatase, Tissue Non-Specific present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Alkaline Phosphatase, Tissue Non-Specific captured in each well. The concentration of Alkaline Phosphatase, Tissue Non-Specific can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.