Specifications
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:PIIINP
- Description:Rat PIIINP ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Rat N-Terminal Propeptide of Collagen alpha-1(III) Chain/PIIINP ELISA Kit exhibits high specificity and excellent specificity for the detection of rat N-Terminal Propeptide of Collagen alpha-1(III) Chain/PIIINP. No significant cross-reactivity or interference between N-Terminal Propeptide of Collagen alpha-1(III) Chain/PIIINP and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.313 - 20 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.188 ng/ml
- Regulatory Status:RUO
- Cat. No.:76739-324
Specifications
About this item
Rat N-Terminal Propeptide of Collagen alpha-1(III) Chain/PIIINP ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat N-Terminal Propeptide of Collagen alpha-1(III) Chain/PIIINP in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 0.313 to 20 ng/ml
- Sensitivity: 0.188 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Rat N-Terminal Propeptide of Collagen alpha-1(III) Chain / PIIINP ELISA Kit (A78628) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat N-Terminal Propeptide of Collagen alpha-1(III) Chain / PIIINP in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for N-Terminal Propeptide of Collagen alpha-1(III) Chain / PIIINP has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the N-Terminal Propeptide of Collagen alpha-1(III) Chain / PIIINP present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-N-Terminal Propeptide of Collagen alpha-1(III) Chain / PIIINP Antibody, which binds the captured N-Terminal Propeptide of Collagen alpha-1(III) Chain / PIIINP present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of N-Terminal Propeptide of Collagen alpha-1(III) Chain / PIIINP captured in each well. The concentration of N-Terminal Propeptide of Collagen alpha-1(III) Chain / PIIINP can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.