- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Thioredoxin Binding Protein 2
- Description:Mouse thioredoxin binding protein 2 ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Mouse Thioredoxin Binding Protein 2 ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Thioredoxin Binding Protein 2. No significant cross-reactivity or interference between Thioredoxin Binding Protein 2 and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.313 - 20 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.188 ng/ml
- Regulatory Status:RUO
- Cat. No.:76738-394
Mouse Thioredoxin Binding Protein 2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Thioredoxin Binding Protein 2 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA kit
- Detection Range: 0.313 to 20 ng/ml
- Sensitivity: 0.188 ng/ml
- Sample Type: Serum, plasma, tissue homogenates and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Mouse Thioredoxin Binding Protein 2 ELISA kit (A77391) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Thioredoxin Binding Protein 2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Thioredoxin Binding Protein 2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Thioredoxin Binding Protein 2 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Thioredoxin Binding Protein 2 Antibody, which binds the captured Thioredoxin Binding Protein 2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Thioredoxin Binding Protein 2 captured in each well. The concentration of Thioredoxin Binding Protein 2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.