- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:TATA binding protein TBP
- Description:Human TATA binding protein TBP ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Human TATA binding protein TBP ELISA Kit exhibits high specificity and excellent specificity for the detection of human TATA binding protein TBP. No significant cross-reactivity or interference between TATA binding protein TBP and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.313 - 20 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.188 ng/ml
- Regulatory Status:RUO
- Cat. No.:76735-884
Human TATA binding protein TBP ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TATA binding protein TBP in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA kit
- Detection Range: 0.313 to 20 ng/ml
- Sensitivity: 0.188 ng/ml
- Sample Type: Serum, plasma, tissue homogenates and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Human TATA binding protein TBP ELISA kit (A78844) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human TATA binding protein TBP in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for TATA binding protein TBP has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the TATA binding protein TBP present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-TATA binding protein TBP Antibody, which binds the captured TATA binding protein TBP present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of TATA binding protein TBP captured in each well. The concentration of TATA binding protein TBP can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.