Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Kidney Androgen-Regulated Protein/KAP ELISA Kit
Mouse Kidney Androgen-Regulated Protein/KAP ELISA Kit
Catalog # 76735-664
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse Kidney Androgen-Regulated Protein/KAP ELISA Kit
Catalog # 76735-664
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A79487-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Kidney Androgen-Regulated Protein/KAP
  • Description:
    Mouse kidney androgen-regulated protein/KAP ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Mouse Kidney Androgen-Regulated Protein/KAP ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Kidney Androgen-Regulated Protein/KAP. No significant cross-reactivity or interference between Kidney Androgen-Regulated Protein/KAP and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    50 μl
  • Sensitivity:
    0.094 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76735-664

Specifications

About this item

Mouse Kidney Androgen-Regulated Protein/KAP ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of mouse Kidney Androgen-Regulated Protein/KAP in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 0.156 to 10 ng/ml
  • Sensitivity: 0.094 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Mouse Kidney Androgen-Regulated Protein/KAP ELISA kit (A79487) employs the competitive enzyme immunoassay technique for the quantitative measurement of mouse Kidney Androgen-Regulated Protein/KAP in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Kidney Androgen-Regulated Protein/KAP antigen. During the incubation, Kidney Androgen-Regulated Protein/KAP present in the samples or standards competes with the fixed amount of immobilized Kidney Androgen-Regulated Protein/KAP for binding sites on the Biotinylated Anti-Kidney Androgen-Regulated Protein/KAP Antibody. The more Kidney Androgen-Regulated Protein/KAP present in a sample or standard, the less Biotinylated Anti-Kidney Androgen-Regulated Protein/KAP Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Kidney Androgen-Regulated Protein/KAP Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Kidney Androgen-Regulated Protein/KAP present in each sample or standard. The concentration of Kidney Androgen-Regulated Protein/KAP can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.