Order Entry
Puerto Rico
ContactUsLinkComponent
Human Free Triiodothyronine/T3 ELISA Kit
Human Free Triiodothyronine/T3 ELISA Kit
Catalog # 76734-244
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human Free Triiodothyronine/T3 ELISA Kit
Catalog # 76734-244
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A78109-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Free Triiodothyronine/T3
  • Description:
    Human free triiodothyronine/T3 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Human Free Triiodothyronine/T3 ELISA Kit exhibits high specificity and excellent specificity for the detection of human Free Triiodothyronine/T3. No significant cross-reactivity or interference between Free Triiodothyronine/T3 and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.781 - 50 pg/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    50 μl
  • Sensitivity:
    0.469 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76734-244

Specifications

About this item

Human Free Triiodothyronine/T3 ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of human Free Triiodothyronine/T3 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 0.781 to 50 pg/ml
  • Sensitivity: 0.469 pg/ml
  • Sample type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human Free Triiodothyronine/T3 ELISA kit (A78109) employs the competitive enzyme immunoassay technique for the quantitative measurement of human Free Triiodothyronine/T3 in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Free Triiodothyronine/T3 antigen. During the incubation, Free Triiodothyronine/T3 present in the samples or standards competes with the fixed amount of immobilized Free Triiodothyronine/T3 for binding sites on the Biotinylated Anti-Free Triiodothyronine/T3 Antibody. The more Free Triiodothyronine/T3 present in a sample or standard, the less Biotinylated Anti-Free Triiodothyronine/T3 Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Free Triiodothyronine/T3 Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Free Triiodothyronine/T3 present in each sample or standard. The concentration of Free Triiodothyronine/T3 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.