- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Lumican
- Description:Mouse lumican ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross Reactivity:Mouse Lumican ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Lumican. No significant cross-reactivity or interference between Lumican and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.625 - 40 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.375 ng/ml
- Regulatory Status:RUO
- Cat. No.:76731-984
Mouse Lumican ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Lumican in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Mouse Lumican ELISA kit (A247343) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Lumican in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Lumican has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Lumican present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Lumican Antibody, which binds the captured Lumican present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Lumican captured in each well. The concentration of Lumican can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.