Order Entry
Puerto Rico
ContactUsLinkComponent
Rat CSNK2A2 ELISA Kit
Rat CSNK2A2 ELISA Kit
Catalog # 76731-794
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Rat CSNK2A2 ELISA Kit
Catalog # 76731-794
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A76328-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    CSNK2A2
  • Description:
    Rat CSNK2A2 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Rat CSNK2A2 ELISA Kit exhibits high specificity and excellent specificity for the detection of rat CSNK2A2. No significant cross-reactivity or interference between CSNK2A2 and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    78.125 - 5000 pg/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    46.875 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76731-794

Specifications

About this item

Rat CSNK2A2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat CSNK2A2 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 78.125 to 5000 pg/ml
  • Sensitivity: 46.875 pg/ml
  • Sample type: Serum. plasma. tissue homogenates and other biological fluids
  • Assay precision: Intra assay: CV <8%. Inter assay: CV <10%

Rat CSNK2A2 ELISA kit (A76328) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat CSNK2A2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for CSNK2A2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the CSNK2A2 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-CSNK2A2 Antibody, which binds the captured CSNK2A2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of CSNK2A2 captured in each well. The concentration of CSNK2A2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.