Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Carbonic Anhydrase 2/CA2 ELISA Kit
Mouse Carbonic Anhydrase 2/CA2 ELISA Kit
Catalog # 76731-104
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse Carbonic Anhydrase 2/CA2 ELISA Kit
Catalog # 76731-104
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A77788-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Carbonic Anhydrase 2/CA2
  • Description:
    Mouse carbonic anhydrase 2/CA2 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Mouse Carbonic Anhydrase 2/CA2 ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Carbonic Anhydrase 2/CA2. No significant cross-reactivity or interference between Carbonic Anhydrase 2/CA2 and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    1.563 - 100 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.938 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76731-104

Specifications

About this item

Mouse Carbonic Anhydrase 2/CA2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Carbonic Anhydrase 2/CA2 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 1.563 to 100 ng/ml
  • Sensitivity: 0.938 ng/ml
  • Sample type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse Carbonic Anhydrase 2/CA2 ELISA kit (A77788) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Carbonic Anhydrase 2/CA2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Carbonic Anhydrase 2/CA2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Carbonic Anhydrase 2/CA2 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Carbonic Anhydrase 2/CA2 Antibody, which binds the captured Carbonic Anhydrase 2/CA2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Carbonic Anhydrase 2/CA2 captured in each well. The concentration of Carbonic Anhydrase 2/CA2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.