- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Other
- Target protein:Claudin 7/CLDN7
- Description:Porcine claudin 7/CLDN7 ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Porcine Claudin 7/CLDN7 ELISA kit exhibits high specificity and excellent specificity for the detection of porcine Claudin 7/CLDN7. No significant cross-reactivity or interference between Claudin 7/CLDN7 and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:125 - 8000 pg/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:75 pg/ml
- Regulatory Status:RUO
- Cat. No.:76747-418
Porcine Claudin 7/CLDN7 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of porcine Claudin 7/CLDN7 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 125 - 8000 pg/ml
- Sensitivity: 75 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Porcine Claudin 7/CLDN7 ELISA kit (A75093) employs the sandwich enzyme immunoassay technique for the quantitative measurement of porcine Claudin 7/CLDN7 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Claudin 7/CLDN7 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Claudin 7/CLDN7 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Claudin 7/CLDN7 Antibody, which binds the captured Claudin 7/CLDN7 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Claudin 7/CLDN7 captured in each well. The concentration of Claudin 7/CLDN7 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.