- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Complement C4a
- Description:Mouse complement C4a ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Mouse Complement C4a ELISA kit exhibits high specificity and excellent specificity for the detection of mouse Complement C4a. No significant cross-reactivity or interference between Complement C4a and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:78.125 - 5000 pg/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:46.875 pg/ml
- Regulatory Status:RUO
- Cat. No.:76746-978
Mouse Complement C4a ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Complement C4a in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 78.125 to 5000 pg/ml
- Sensitivity: 46.875 pg/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse Complement C4a ELISA kit (A77777) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Complement C4a in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Complement C4a has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Complement C4a present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Complement C4a Antibody, which binds the captured Complement C4a present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Complement C4a captured in each well. The concentration of Complement C4a can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.