- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:COMP
- Description:Rat Comp ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Rat COMP/Cartilage Oligomeric Matrix Protein ELISA Kit exhibits high specificity and excellent specificity for the detection of rat COMP/Cartilage Oligomeric Matrix Protein. No significant cross-reactivity or interference between COMP/Cartilage Oligomeric Matrix Protein and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:1.563 - 100 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.938 ng/ml
- Regulatory Status:RUO
- Cat. No.:76746-898
Rat COMP/Cartilage Oligomeric Matrix Protein ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat COMP/Cartilage Oligomeric Matrix Protein in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 1.563 to 100 ng/ml
- Sensitivity: 0.938 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Rat COMP / Cartilage Oligomeric Matrix Protein ELISA Kit (A79941) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat COMP / Cartilage Oligomeric Matrix Protein in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for COMP / Cartilage Oligomeric Matrix Protein has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the COMP / Cartilage Oligomeric Matrix Protein present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-COMP / Cartilage Oligomeric Matrix Protein Antibody, which binds the captured COMP / Cartilage Oligomeric Matrix Protein present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of COMP / Cartilage Oligomeric Matrix Protein captured in each well. The concentration of COMP / Cartilage Oligomeric Matrix Protein can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.